ContactUsLinkComponent
Recombinant protein, gyg2( aah23152.1, 1 a.a. - 470 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008908-P01
Supplier:  Abnova
Recombinant protein, gyg2( aah23152.1, 1 a.a. - 470 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008908-P01
Supplier:  Abnova