ContactUsLinkComponent
Recombinant protein, tsn( np_004613.1, 1 a.a. - 228 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007247-P02
Supplier:  Abnova
Recombinant protein, tsn( np_004613.1, 1 a.a. - 228 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007247-P02
Supplier:  Abnova