ContactUsLinkComponent
Recombinant protein, trim28( np_005753.1, 1 a.a. - 835 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010155-P01
Supplier:  Abnova
Recombinant protein, trim28( np_005753.1, 1 a.a. - 835 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010155-P01
Supplier:  Abnova
Frequently Bought Together