ContactUsLinkComponent
Recombinant protein, snx3( aah16863, 1 a.a. - 114 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008724-P01
Supplier:  Abnova
Recombinant protein, snx3( aah16863, 1 a.a. - 114 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00008724-P01
Supplier:  Abnova