ContactUsLinkComponent
Recombinant protein, cdc23( aah17713, 1 a.a. - 592 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00008697-P01
Supplier:  Abnova
undefined
Recombinant protein, cdc23( aah17713, 1 a.a. - 592 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00008697-P01
Supplier:  Abnova
Frequently Bought Together