ContactUsLinkComponent
SOLUTION MULTIELEMENTAIRE DE 21 COMPOSES DANS HNO3 5% Al 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Mg 50mg/l Mo 50mg/l Ni 50mg/l Pb 50mg/l Zn 50mg/l Sn 50mg/l V 50mg/l Ba 50mg/l Fe 50mg/l Ca 50mg/l Sb 50mg/l As 200mg/l 200m 1 * 250 mL
Catalog # CMPL047B.50.5N.L25
SOLUTION MULTIELEMENTAIRE DE 21 COMPOSES DANS HNO3 5% Al 50mg/l Cd 50mg/l Co 50mg/l Cr 50mg/l Mg 50mg/l Mo 50mg/l Ni 50mg/l Pb 50mg/l Zn 50mg/l Sn 50mg/l V 50mg/l Ba 50mg/l Fe 50mg/l Ca 50mg/l Sb 50mg/l As 200mg/l 200m 1 * 250 mL
Catalog # CMPL047B.50.5N.L25
Frequently Bought Together