ContactUsLinkComponent
Recombinant protein, saps2( np_055493.2, 1 a.a. - 932 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009701-P01
Supplier:  Abnova
Recombinant protein, saps2( np_055493.2, 1 a.a. - 932 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009701-P01
Supplier:  Abnova