ContactUsLinkComponent
Recombinant protein, ube3c( aah26241.1, 1 a.a. - 404 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009690-P01
Supplier:  Abnova
Recombinant protein, ube3c( aah26241.1, 1 a.a. - 404 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009690-P01
Supplier:  Abnova