ContactUsLinkComponent
Recombinant protein, zp2( aah96305.1, 1 a.a. - 736 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007783-P01
Supplier:  Abnova
Recombinant protein, zp2( aah96305.1, 1 a.a. - 736 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00007783-P01
Supplier:  Abnova