ContactUsLinkComponent
Recombinant protein, bst2( aah33873.1, 1 a.a. - 181 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000684-P02
Supplier:  Abnova
Recombinant protein, bst2( aah33873.1, 1 a.a. - 181 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000684-P02
Supplier:  Abnova