ContactUsLinkComponent
Recombinant protein, bmp7( aah08584, 1 a.a. - 432 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000655-P01
Supplier:  Abnova
Recombinant protein, bmp7( aah08584, 1 a.a. - 432 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000655-P01
Supplier:  Abnova