ContactUsLinkComponent
Recombinant protein, glo1( aah11365, 1 a.a. - 185 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002739-P01
Supplier:  Abnova
Recombinant protein, glo1( aah11365, 1 a.a. - 185 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002739-P01
Supplier:  Abnova