ContactUsLinkComponent
Recombinant protein, ndufb6( aah29247, 1 a.a. - 129 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004712-P01
Supplier:  Abnova
Recombinant protein, ndufb6( aah29247, 1 a.a. - 129 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00004712-P01
Supplier:  Abnova