ContactUsLinkComponent
Recombinant protein, gak( aah08668, 24 a.a. - 417 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
  ABNOH00002580-P01
 :  Abnova
Recombinant protein, gak( aah08668, 24 a.a. - 417 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
  ABNOH00002580-P01
 :  Abnova
Frequently Bought Together