ContactUsLinkComponent
Recombinant protein, ap1b1( cak54367.1, 1 a.a. - 939 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000162-P01
Supplier:  Abnova
Recombinant protein, ap1b1( cak54367.1, 1 a.a. - 939 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000162-P01
Supplier:  Abnova