ContactUsLinkComponent
Recombinant protein, epha7( aah27940.1, 1 a.a. - 279 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002045-P01
Supplier:  Abnova
Recombinant protein, epha7( aah27940.1, 1 a.a. - 279 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00002045-P01
Supplier:  Abnova