ContactUsLinkComponent
Recombinant protein, pou3f2( aah51699.2, 1 a.a. - 443 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005454-P01
Supplier:  Abnova
Recombinant protein, pou3f2( aah51699.2, 1 a.a. - 443 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005454-P01
Supplier:  Abnova