Recombinant protein, eif2b1( np_001405.1, 1 a.a. - 305 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001967-P01
Dodavatel: Abnova
Recombinant protein, eif2b1( np_001405.1, 1 a.a. - 305 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00001967-P01
Dodavatel: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR® Facial Tissues
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR® Serological Pipette, Standard Line
Dodavatel: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Dodavatel: VWR Collection