ContactUsLinkComponent
62 µL large aperture Transfer Pipettes, Standard. Length 13,7 cm. Stem Diameter 6,1 mm. Overall Capacity 9,5 mL. Bulb Draw 6,2 mL. 20 drops per mL. 1 * 2.500 items
Catalog # SAMC691
Supplier:  SAMCO
undefined
62 µL large aperture Transfer Pipettes, Standard. Length 13,7 cm. Stem Diameter 6,1 mm. Overall Capacity 9,5 mL. Bulb Draw 6,2 mL. 20 drops per mL. 1 * 2.500 items
Catalog # SAMC691
Supplier:  SAMCO
Frequently Bought Together