ContactUsLinkComponent
Transfer Pipettes, 6 B/B Pet (Blood Bank). Length 15,4 cm. Stem Diameter 6,6 mm. Overall Capacity 4,9 mL. Bulb Draw 1,8 mL. 24 drops per mL. 1 * 500 items
Catalog # SAMC336
Supplier:  SAMCO
undefined
Transfer Pipettes, 6 B/B Pet (Blood Bank). Length 15,4 cm. Stem Diameter 6,6 mm. Overall Capacity 4,9 mL. Bulb Draw 1,8 mL. 24 drops per mL. 1 * 500 items
Catalog # SAMC336
Supplier:  SAMCO