ContactUsLinkComponent
30 COMPOSES DANS HNO3 5%,HF 0,5% (prepared from salts with high purity 99,995+%) Al 1mg/l As 1mg/l B 1mg/l Ba 1mg/l Bi 1mg/l Ca 1mg/l Cd 1mg/l Co 1mg/l Cr 1mg/l Cu 1mg/l Fe 1mg/l Hg 1mg/l K 1mg/l Mg 1mg/l Mn 1mg/l Mo P 1 * 500 mL
Catalog # CMPLE200.1.5N05F
30 COMPOSES DANS HNO3 5%,HF 0,5% (prepared from salts with high purity 99,995+%) Al 1mg/l As 1mg/l B 1mg/l Ba 1mg/l Bi 1mg/l Ca 1mg/l Cd 1mg/l Co 1mg/l Cr 1mg/l Cu 1mg/l Fe 1mg/l Hg 1mg/l K 1mg/l Mg 1mg/l Mn 1mg/l Mo P 1 * 500 mL
Catalog # CMPLE200.1.5N05F
Frequently Bought Together