ContactUsLinkComponent
SOLUTION COV 21 COMPOSES DANS METHANOL (Ampoules de 1 ml ) Vinylchloride [CAS:75-01-4] 25mg/l Dichloromethane [CAS:75-09-2] 250mg/l Chloroform [CAS:67-66-3] 25mg/l 1,1,1-Trichloroethane [CAS:71-55-6] 10mg/l Tetrachloromethane [CAS:56-23-5] ,2 1 * 10 Ampoul
Katalogové číslo CMPL767E.50.ML.1
SOLUTION COV 21 COMPOSES DANS METHANOL (Ampoules de 1 ml ) Vinylchloride [CAS:75-01-4] 25mg/l Dichloromethane [CAS:75-09-2] 250mg/l Chloroform [CAS:67-66-3] 25mg/l 1,1,1-Trichloroethane [CAS:71-55-6] 10mg/l Tetrachloromethane [CAS:56-23-5] ,2 1 * 10 Ampoul
Katalogové číslo CMPL767E.50.ML.1
Frequently Bought Together