ContactUsLinkComponent
19COMPOSES A 100 MG/L CHAQUE DS HNO3 5% of Al As Ag B Ba Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Ti V Zn 1 * 500 mL
Catalog # CMPLFD79.K1.5N.L5
19COMPOSES A 100 MG/L CHAQUE DS HNO3 5% of Al As Ag B Ba Cd Co Cr Cu Fe Mn Mo Ni Pb Sb Se Ti V Zn 1 * 500 mL
Catalog # CMPLFD79.K1.5N.L5
Frequently Bought Together