ContactUsLinkComponent
7 COMPOSES A 10 MG/L CHAQUE DANS H2O (AMPOULE DE 10 ML ) of Chlorides (Cl-) Fluorides (F-) Nitrates (NO3-) Sulphates (SO42-) Formiates (HCOO-) Acetates (CH3COO-) Phosphates (PO43-) 1 * 1 Ampoul
Catalog # CMPLC54D.10.W.10
7 COMPOSES A 10 MG/L CHAQUE DANS H2O (AMPOULE DE 10 ML ) of Chlorides (Cl-) Fluorides (F-) Nitrates (NO3-) Sulphates (SO42-) Formiates (HCOO-) Acetates (CH3COO-) Phosphates (PO43-) 1 * 1 Ampoul
Catalog # CMPLC54D.10.W.10
Frequently Bought Together