ContactUsLinkComponent
Whirl-Pak® Sample Bag, Sterile, 207 ml, pk/100, size: 9.5 cm x 18 cm (W x L), without Dechlorinating Agent 1 * 100 items
Catalog # HACH2233199
Supplier:  Hach
undefined
Whirl-Pak® Sample Bag, Sterile, 207 ml, pk/100, size: 9.5 cm x 18 cm (W x L), without Dechlorinating Agent 1 * 100 items
Catalog # HACH2233199
Supplier:  Hach
Frequently Bought Together