ContactUsLinkComponent
Interferin-hts sirna transfection reagent is especially developed for high troughput screening (hts) applications providing great si lencing efficiency, excellent reproducibility and high cell viability with very low amounts of reagent. suitable for 1 * 6 ml
Catalog # 410-060
Interferin-hts sirna transfection reagent is especially developed for high troughput screening (hts) applications providing great si lencing efficiency, excellent reproducibility and high cell viability with very low amounts of reagent. suitable for 1 * 6 ml
Catalog # 410-060