ContactUsLinkComponent
Jetpei-huvec is a dna transfection reagent optimized for the transfection of primary human endothelial cells such as huvec (human um bilical vein endothelial cells). also recommended for the transfection of vascular endothelial cells of various origi 1 * 100 µl
Catalog # 108-01N
Jetpei-huvec is a dna transfection reagent optimized for the transfection of primary human endothelial cells such as huvec (human um bilical vein endothelial cells). also recommended for the transfection of vascular endothelial cells of various origi 1 * 100 µl
Catalog # 108-01N