1,10-Fenantroliny chlorowodorek czda >99 % wsk 25g - oświadczenie 1 * 25 g
Catalog # SIAL77510-25G
Supplier: Merck
1,10-Fenantroliny chlorowodorek czda >99 % wsk 25g - oświadczenie 1 * 25 g
Catalog # SIAL77510-25G
Supplier: Merck
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide, Reagent ACS Grade 1 * 500 mL
Supplier: Spectrum Chemical
1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific