ContactUsLinkComponent
Snapmop polylmesh 36cm for use with slimline series adapter. no exposed foam. polymesh material is ideal for removing heavy deposits such as photo resist. can be used on floors, walls and ceilings. compatible with strong disinfectants. launderable, 1 * 24 items
Catalog # MICRMZSM68-14
Supplier:  MICRONOVA
Snapmop polylmesh 36cm for use with slimline series adapter. no exposed foam. polymesh material is ideal for removing heavy deposits such as photo resist. can be used on floors, walls and ceilings. compatible with strong disinfectants. launderable, 1 * 24 items
Catalog # MICRMZSM68-14
Supplier:  MICRONOVA
Frequently Bought Together