ContactUsLinkComponent
14 COMPOSES DS CARBONE DISULFURE LOT A 2000mg/l each of Butane [CAS:106-97-8] Isobutane [CAS:75-28-5] 2-Methylbutane [CAS:78-78-4] n-Pentane [CAS:109-66-0] n-Hexane [CAS:110-54-3] 2-Methylpentane [CAS:107-83-5] n-Heptane [CAS:142-82-5] 1 * 5 Ampoul
Catalog # CMPLEB45.2K.CDS
14 COMPOSES DS CARBONE DISULFURE LOT A 2000mg/l each of Butane [CAS:106-97-8] Isobutane [CAS:75-28-5] 2-Methylbutane [CAS:78-78-4] n-Pentane [CAS:109-66-0] n-Hexane [CAS:110-54-3] 2-Methylpentane [CAS:107-83-5] n-Heptane [CAS:142-82-5] 1 * 5 Ampoul
Catalog # CMPLEB45.2K.CDS
Frequently Bought Together