ContactUsLinkComponent
DRY DISPERSING UNIT f. ANALYSETTE 22 MICRO TEC PLUS Sample volume:,,< 1 cm3 to 100 cm3 Dimensions(W x D x H):,,36 x 65 x 67 cm Mass:,,25 kg 1 * 1 items
Catalog # FRIT22.8600.00
Supplier:  FRITSCH
DRY DISPERSING UNIT f. ANALYSETTE 22 MICRO TEC PLUS Sample volume:,,< 1 cm3 to 100 cm3 Dimensions(W x D x H):,,36 x 65 x 67 cm Mass:,,25 kg 1 * 1 items
Catalog # FRIT22.8600.00
Supplier:  FRITSCH
Frequently Bought Together