ContactUsLinkComponent
Tube ps 16x108 15ml disposablemm,disposable tubes - polystyrene (ps) transparent, very inexpensive. for solvents and temperatures ab ove 70°c 16x108 15ml. 1 * 100 items
Catalog # SEMA1595
Supplier:  SEMADENI
Tube ps 16x108 15ml disposablemm,disposable tubes - polystyrene (ps) transparent, very inexpensive. for solvents and temperatures ab ove 70°c 16x108 15ml. 1 * 100 items
Catalog # SEMA1595
Supplier:  SEMADENI
Frequently Bought Together