ContactUsLinkComponent
gas cylinder trolley with air damped rubber wheels and fold-out tail wheel. for the transport of 2 x 50 litre gas bottles, blue pain ted steel tube construction, inclusive chain restraint. firm grip by hand protection bracket. 1 * 1 items
Catalog # DUEH7.01.040
Supplier:  DUPERTHAL
gas cylinder trolley with air damped rubber wheels and fold-out tail wheel. for the transport of 2 x 50 litre gas bottles, blue pain ted steel tube construction, inclusive chain restraint. firm grip by hand protection bracket. 1 * 1 items
Catalog # DUEH7.01.040
Supplier:  DUPERTHAL
Frequently Bought Together