Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Filter tubes, plain neck

Filter tubes, plain neck

24 mm, 150 mm
Catalog # BIBB2110/08M
Supplier:   SciLabware
Filter tubes, plain neck
Catalog # BIBB2110/08M
Supplier:  SciLabware
Filter tubes, plain neck
Filter tubes, plain neck

24 mm, 150 mm

Price
3 260,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents

These filter tubes can be used for small scale filtrations. They are ideal for use with Pyrex® short stem funnels.

Specifications
  • Tube ext.Ø:
    24 mm
  • Tube length:
    150 mm