ContactUsLinkComponent
Shodex GPC KF-2006M - polymer based organic HPLC column - SEC mode - THF - preparative - ID: 20mm length: 300mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 10 - Pore Size (A): - - Exclusion Limit (g/ 1 * 1 items
Catalog # SHOWF6102409
Supplier:  Resonac
Shodex GPC KF-2006M - polymer based organic HPLC column - SEC mode - THF - preparative - ID: 20mm length: 300mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 10 - Pore Size (A): - - Exclusion Limit (g/ 1 * 1 items
Catalog # SHOWF6102409
Supplier:  Resonac
Frequently Bought Together