COLLAGEN SOLUTION, 6MG/ML, FROM CALF SKIN, STERILE-FILTERED, SUITABLE FOR CELL CULTURE 1 * 50 mL
Catalog # SIALC2124-50ML
Supplier: Merck
COLLAGEN SOLUTION, 6MG/ML, FROM CALF SKIN, STERILE-FILTERED, SUITABLE FOR CELL CULTURE 1 * 50 mL
Catalog # SIALC2124-50ML
Supplier: Merck
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection