ContactUsLinkComponent
Empty Glass GC Column, for Agilent(R) 5880, 5890, 6890, 7820, and 7890 (Configuration &#34 A&#34 ), L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm 1 * 1 Each
Catalog # 21641.
Supplier:  Merck
undefined
Empty Glass GC Column, for Agilent(R) 5880, 5890, 6890, 7820, and 7890 (Configuration &#34 A&#34 ), L x O.D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm 1 * 1 Each
Catalog # 21641.
Supplier:  Merck
Frequently Bought Together