ContactUsLinkComponent
Glass Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase 4% Carbowax 20M, matrix 80/120 Carbopack B DA support, L x O .D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm 1 * 1 Each
Catalog # 25936.
Supplier:  Merck
Glass Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase 4% Carbowax 20M, matrix 80/120 Carbopack B DA support, L x O .D. x I.D. 6.0 ft (1.8 m) x 1/4 in. x 2.0 mm 1 * 1 Each
Catalog # 25936.
Supplier:  Merck
Frequently Bought Together