ContactUsLinkComponent
Precision Sampling syringe needles, volume 100-10000 muL, for use with VICI Series A Syringe, stainless steel needle bevel tip (poin t style 2), needle L x O.D. x I.D. 2.25 in. x 0.029 in. x 0.012 in. 1 * 1 Each
Catalog # SIAL20996
Supplier:  Merck
Precision Sampling syringe needles, volume 100-10000 muL, for use with VICI Series A Syringe, stainless steel needle bevel tip (poin t style 2), needle L x O.D. x I.D. 2.25 in. x 0.029 in. x 0.012 in. 1 * 1 Each
Catalog # SIAL20996
Supplier:  Merck
Frequently Bought Together