ContactUsLinkComponent
Metal Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase 20% TCEP, matrix 80/100 Chromosorb P AW support, L x O.D. x I.D. 1.8 ft (0.55 m) x 1/16 in. x 0.75 mm 1 * 1 Each
Catalog # 12873.
Supplier:  Merck
Metal Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase 20% TCEP, matrix 80/100 Chromosorb P AW support, L x O.D. x I.D. 1.8 ft (0.55 m) x 1/16 in. x 0.75 mm 1 * 1 Each
Catalog # 12873.
Supplier:  Merck
Frequently Bought Together