ContactUsLinkComponent
Metal Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase none, matrix 60/80 Tenax TA support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12554.
Supplier:  Merck
Metal Packed GC Column (fits Agilent, Configuration &#34 A&#34 ), phase none, matrix 60/80 Tenax TA support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12554.
Supplier:  Merck
Frequently Bought Together