Swagelok(R) PTFE Ferrules, PTFE back ferrule, Swagelok for 1/8 in. tubing 1 * 1 Each
Catalog # SIAL22059
Supplier: Merck
Swagelok(R) PTFE Ferrules, PTFE back ferrule, Swagelok for 1/8 in. tubing 1 * 1 Each
Catalog # SIAL22059
Supplier: Merck
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Incinerating dishes
Supplier: MORGAN ADVANCED MATERIALS HALD
Tubing, PTFE, BOLA
Supplier: Bohlender
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals