ContactUsLinkComponent
Solution Etalon multi 5 éléments CI Anions 250ml : 5 components Chlorides (Cl-) 200mg/l Fluorides (F-) 100mg/l Nitrites (NO2-) 500mg/l Nitrates (NO3-) 1000mg/l Sulphates (SO42-) 1000mg/l in Water . 1 * 250 mL
Catalog # CMPLDA6D.K2.W.L25
Solution Etalon multi 5 éléments CI Anions 250ml : 5 components Chlorides (Cl-) 200mg/l Fluorides (F-) 100mg/l Nitrites (NO2-) 500mg/l Nitrates (NO3-) 1000mg/l Sulphates (SO42-) 1000mg/l in Water . 1 * 250 mL
Catalog # CMPLDA6D.K2.W.L25
Frequently Bought Together