8 COMPOSES A 10 MG/L CHAQUE DS H2O POUR IC of Fluorides (F-) Bromides (Br-) Chlorides (Cl-) Sulphates (SO42-) Nitrates (NO3-) Nitrites (NO2-) Phosphates (PO43-) Carbonates (CO32-) 1 * 100 ml
Catalog # CMPLE92A.10.W.L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together