ContactUsLinkComponent
Stopper,replacement,size : NS 45/40,made of glass-fibre reinforced PTFE,for locking and unlocking inner ground joints,SAB. 1 * 1 items
Catalog # BOHLK1349-16
Supplier:  Bohlender
undefined
Stopper,replacement,size : NS 45/40,made of glass-fibre reinforced PTFE,for locking and unlocking inner ground joints,SAB. 1 * 1 items
Catalog # BOHLK1349-16
Supplier:  Bohlender
Frequently Bought Together