ContactUsLinkComponent
Screw fitting,swiveling,for sensor/tube outer diameter: 9.52mm,for thread : GL 25, maximum angle : 9°, fix all kinds of sensors (sta inless steel, glass, PTFE), tubes and tight tubing in the requested angle,suitable for both pressure and vacuum. 1 * 1 items
Catalog # BOHLD690-42
Supplier:  Bohlender
undefined
Screw fitting,swiveling,for sensor/tube outer diameter: 9.52mm,for thread : GL 25, maximum angle : 9°, fix all kinds of sensors (sta inless steel, glass, PTFE), tubes and tight tubing in the requested angle,suitable for both pressure and vacuum. 1 * 1 items
Catalog # BOHLD690-42
Supplier:  Bohlender
Frequently Bought Together