Screw fitting,swiveling,for sensor/tube outer diameter: 19mm,for thread : GL 32, maximum angle : 3°, fix all kinds of sensors (stain less steel, glass, PTFE), tubes and tight tubing in the requested angle,suitable for both pressure and vacuum. 1 * 1 items
Catalog # BOHLD690-68
Supplier: Bohlender
Frequently Bought Together