Orders LinkContactUsLinkComponent
VWR® Basic Beard Covers, SPP
VWR® Basic Beard Covers, SPP
Latex-free, White
Catalog # 113-1464
Supplier:   VWR Collection
VWR® Basic Beard Covers, SPP
Catalog # 113-1464
Supplier:  VWR Collection
VWR® Basic Beard Covers, SPP
VWR® Basic Beard Covers, SPP
Latex-free, White
Price
2 010,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
VWR® Basic SPP beard covers are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility, lot-controlled and validated through independent laboratory testing.
Diameter (stretched): 406 mm
Specifications
  • Colour:
    White
  • Description:
    Latex-free
  • Seq:
    02