Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
VWR® Basic Lab Coats, SMS, Disposable

VWR® Basic Lab Coats, SMS, Disposable

White, S, 93,35 cm
Catalog # 113-1131
Supplier:   VWR Collection
VWR® Basic Lab Coats, SMS, Disposable
Catalog # 113-1131
Supplier:  VWR Collection
VWR® Basic Lab Coats, SMS, Disposable
VWR® Basic Lab Coats, SMS, Disposable

White, S, 93,35 cm

Price
4 830,00 Kč
Log in for availability
1 * 30 KS
Product Details & Documents

SMS lab coats are manufactured from a latex-free, multi-layer spunbonded-melt-blown-spunbonded (SMS) fabric that offers high tensile strength, while remaining soft, comfortable and breathable.

  • Tapered collar, inset sleeves and snap closure
  • Knitted cuffs and three pockets
  • Fluid and particulate barrier
  • Light chemical splash resistance

Colour: White or blue

Specifications
  • Clothing Size:
    S
  • Colour:
    White
  • Length:
    93,35 cm
  • Seq:
    01
  • Cat. No.:
    113-1131
  • Garment application:
    Cleanroom|Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.