ContactUsLinkComponent
The maximum speed for this tool should not exceed 24'000 rpm although the drive is able to run at 30'000rpm 1 * 1 items
Catalog # KINE11030254
Supplier:  KINEMATICA
The maximum speed for this tool should not exceed 24'000 rpm although the drive is able to run at 30'000rpm 1 * 1 items
Catalog # KINE11030254
Supplier:  KINEMATICA
Frequently Bought Together